Learn More
Innovative Research Inc FXI Dfcnt Hmn Pla 1ml
Catalog No. 502744591
Encompass_Preferred
Factor XI (FXI) Deficient Human Plasma 1ml - Factor XI (FXI) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor XI so that the remaining activity is less than or equal to 1%.
Catalog No. 50-274-4591 Supplier Innovative Research Inc Supplier No. IFXIDP1ML
Factor XI (FXI) Deficient Human Plasma 1ml - Factor XI (FXI) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor XI so that the remaining activity is less than or equal to 1%.