Learn More
Innovative Research Inc FXII Dfcnt Hmn Pla 50ml
Catalog No. 502744589
Encompass_Preferred
Factor XII (FXII) Deficient Human Plasma 50ml - Factor XII (FXII) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor XII so that the remaining activity is less than or equal to 1%.
Catalog No. 50-274-4589 Supplier Innovative Research Inc Supplier No. IFXIIDP50ML
Factor XII (FXII) Deficient Human Plasma 50ml - Factor XII (FXII) Deficient Human Plasma from Innovative Research is prepared from high-quality, citrated pooled human plasma that has been immuno-depleted of Factor XII so that the remaining activity is less than or equal to 1%.