Learn More
ABclonal Technology FXR/NR1H4 Rabbit mAb
SDP
Catalog No. 5000007345
Encompass_Preferred
This gene encodes a ligand-activated transcription factor that shares structural features in common with nuclear hormone receptor family members This protein functions as a receptor for bile acids and when bound to bile acids binds to DNA and regulates the expression of genes involved in bile acid synthesis and transport Alternatively spliced transcript variants encoding different isoforms have been described
Catalog No. 50-000-07345 Supplier ABclonal Technology Supplier No. A24015100UL
This gene encodes a ligand-activated transcription factor that shares structural features in common with nuclear hormone receptor family members This protein functions as a receptor for bile acids and when bound to bile acids binds to DNA and regulates the expression of genes involved in bile acid synthesis and transport Alternatively spliced transcript variants encoding different isoforms have been described