Learn More
Aviva Systems Biology Corp FYB1 Antibody - N-terminal region (ARP63469_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63469_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FYB. It was validated on Western Blot. . Alternate names: ADAP, PRO0823, SLAP-130ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-287-978