Learn More
Aviva Systems Biology Corp FYB1 Antibody - C-terminal region (ARP63470_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63470_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FYB. It was validated on Western Blot. . Alternate names: ADAP, PRO0823, SLAP-130ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse
Catalog No. 50-287-979