Learn More
ABclonal Technology FYB Rabbit pAb
SDP
Supplier:  ABclonal Technology A159971000UL
Encompass_Preferred
The protein encoded by this gene is an adapter for the FYN protein and LCP2 signaling cascades in T-cells. The encoded protein is involved in platelet activation and controls the expression of interleukin-2. Three transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-06536