Learn More
Aviva Systems Biology Corp FYN Antibody - middle region (ARP55568_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55568_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FYN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FYN. Alternate names: MGC45350, SLK, SYN, p59-FYNApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-63020