Learn More
Enzo Life Sciences Fyn kinase substrate (1mg)
SDP
Catalog No. 502011290
Encompass_Preferred
This peptide, EFGTYGTLSKKK, is derived from a peptide (pF4) discovered, through phage display, to be a very good substrate for Fyn kinase. Its Km is estimated to be 70 µM. The addition of three lysines on the C-terminus allows this peptide to be used in phosphocellulose kinase assays. Purity: ≥95%. Long Term Storage: -20°C.
Catalog No. 50-201-1290 Supplier Enzo Life Sciences Supplier No. BMLP2150001
This peptide, EFGTYGTLSKKK, is derived from a peptide (pF4) discovered, through phage display, to be a very good substrate for Fyn kinase. Its Km is estimated to be 70 µM. The addition of three lysines on the C-terminus allows this peptide to be used in phosphocellulose kinase assays. Purity: ≥95%. Long Term Storage: -20°C.