Learn More
Pyramex™ Fyxate™ Safety Eyewear
Catalog No. 19155640 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Indoor/Outdoor, Mirror
Description:
Dielectric
Dielectric with Foam padding and quick release strap
Lens Coating:
Anti-fog
Uncoated
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Description Lens Coating
19-155-640 Indoor/Outdoor, Mirror Dielectric -
19-155-634 Clear Dielectric Uncoated
19-155-635 Clear Dielectric Anti-fog
19-155-636 Clear Dielectric with Foam padding and quick release strap Anti-fog
19-155-637 Gray Dielectric Uncoated
19-155-638 Gray Dielectric Anti-fog
19-155-639 Gray Dielectric with Foam padding and quick release strap Anti-fog
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-155-640 Supplier Pyramex™ Supplier No. SB10280S
Add to Cart
Add to Cart
This item is not returnable. View return policy
Dielectric polycarbonate safety glasses
  • Dielectric
  • Rubber temple tips and nosepiece
  • Single wraparound lens
  • Specifications
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Description Dielectric
    Lens Tint Indoor/Outdoor, Mirror
    Frame Color Black
    Lens Material Polycarbonate
    Frame Material Polycarbonate