Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for Large 3.75" High Boxes, 3 x 2 Array
Catalog No. 501960817 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer and then sliding the boxes out from either side of the rack.
Catalog No. 50-196-0817 Supplier Cole-Parmer Supplier No. 0440464
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer and then sliding the boxes out from either side of the rack.