Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for Microtube Boxes with 2 7/16" Max Height, 5 x 3 Array
Catalog No. 501960771 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks have drawers that slide out for quick access and storage of boxes. Boxes can be accessed from either side of the rack. They are crafted of corrosion-resist stainless steel for long-term use.
Catalog No. 50-196-0771 Supplier Cole-Parmer Supplier No. 0440212
These upright freezer racks have drawers that slide out for quick access and storage of boxes. Boxes can be accessed from either side of the rack. They are crafted of corrosion-resist stainless steel for long-term use.