Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for Microtube Boxes with 2 7/16" Max Height, 4 x 4 Array
Catalog No. 501960523 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer drawer racks features drawers that slide out for quick access and storage of sample tubes. They are crafted of corrosion-resistent stainless steel for durablity.
Catalog No. 50-196-0523 Supplier Cole-Parmer Supplier No. 0440210
These upright freezer drawer racks features drawers that slide out for quick access and storage of sample tubes. They are crafted of corrosion-resistent stainless steel for durablity.