Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for 25-Place Slide Boxes, 6 x 7 Array
Catalog No. 501960781 Shop All Cole-Parmer Products
Encompass_Preferred
These upright freezer racks have drawers that slide out for quick access and storage of boxes. Built-in label holder lets you conveniently identify each drawer content. Boxes can be accessed from either side of the rack. They are crafted of corrosion-resist stainless steel for long-term use.
Catalog No. 50-196-0781 Supplier Cole-Parmer Supplier No. 0440271
These upright freezer racks have drawers that slide out for quick access and storage of boxes. Built-in label holder lets you conveniently identify each drawer content. Boxes can be accessed from either side of the rack. They are crafted of corrosion-resist stainless steel for long-term use.