Learn More
Cole-Parmer Argos Technologies PolarSafe Adjustable Upright Freezer Rack for Standard 2" & 3" Boxes; holds (15) 2", (6) 3", or (9) 2"+(3) 3" boxes
Catalog No. 501960548 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Adjustable clips offer the flexibility to hold 2"or 3" boxes or a combination of 2" and 3" boxes.
Catalog No. 50-196-0548 Supplier Cole-Parmer Supplier No. 0440479
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Adjustable clips offer the flexibility to hold 2"or 3" boxes or a combination of 2" and 3" boxes.