Learn More
Cole-Parmer Argos Technologies PolarSafe Adjustable Upright Freezer Rack for Standard 2" & 3" Boxes; holds (16) 2", (8) 3", or (8) 2"+(4) 3" boxes
Catalog No. 501960549 Shop All Cole-Parmer Products
Encompass_Preferred
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Adjustable clips offer the flexibility to hold 2"or 3" boxes or a combination of 2" and 3" boxes.
Catalog No. 50-196-0549 Supplier Cole-Parmer Supplier No. 0440481
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Adjustable clips offer the flexibility to hold 2"or 3" boxes or a combination of 2" and 3" boxes.