Learn More
Cayman Chemical Fz721trIfluoroACTAte salt 10mg
SDP
Catalog No. 502915222
Encompass_Preferred
A peptide antagonist of the FZD7 receptor (IC50 = 100 nM); inhibits Wnt3a-induced β-catenin signaling in a reporter assay using HEK293 cells; reduces the number of Lrg5-positive cells and the mRNA encoding Ash-2 and Axin2 and increases mRNA encoding Muc2 in primary mouse intestinal organoids at 100 µM
Catalog No. 50-291-5222 Supplier Cayman Chemical Supplier No. 3692210 MG
A peptide antagonist of the FZD7 receptor (IC50 = 100 nM); inhibits Wnt3a-induced β-catenin signaling in a reporter assay using HEK293 cells; reduces the number of Lrg5-positive cells and the mRNA encoding Ash-2 and Axin2 and increases mRNA encoding Muc2 in primary mouse intestinal organoids at 100 µM