Learn More
Aviva Systems Biology Corp FZD10 Antibody - N-terminal region (ARP41262_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41262_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FZD10. Alternate names: FZ-10, FzE7, hFz10, Fz10, CD350ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Zebrafish
Catalog No. 50-014-57784