Learn More
Aviva Systems Biology Corp FZD2 Antibody - N-terminal region (ARP41236_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41236_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FZD2. Alternate names: Fz2, fz-2, fzE2, hFz2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rat
Catalog No. 50-014-57774