Learn More
Aviva Systems Biology Corp FZD5 Antibody - middle region (ARP41244_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41244_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD5. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FZD5. Alternate names: C2orf31, DKFZP434E2135, HFZ5, MGC129692ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57778