Learn More
Aviva Systems Biology Corp FZD6 Antibody - middle region (ARP41249_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41249_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human FZD6. Alternate names: Hfz6, FZ6, FZ-6, HFZ6, NDNC10ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57780