Learn More
Aviva Systems Biology Corp FZD9 Antibody - N-terminal region (ARP41253_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41253_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FZD9. Alternate names: FZD3, CD349ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57783