Learn More
Aviva Systems Biology Corp FZD9 Antibody - N-terminal region (ARP41252_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41252_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZD9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human FZD9. Alternate names: FZD3, CD349ApplicationWB. Species reactivity: Dog, Guinea Pig, Human, Mouse, Rat
Catalog No. 50-014-57782