Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Drawer Freezer Rack for 0.2 mL Tube 96-Well PCR Boxes, 3 x 2 x 2 Array
Catalog No. 501960829 Shop All Cole-Parmer Products
Encompass_Preferred
Each compartment of this drawer rack holds two, 96-well boxes that easily can be retrieved from either side of rack. It is made of sturdy corrosion-resistant stainless steel for long-term use. Drawers slide out independently for quick access and storage of boxes.
Catalog No. 50-196-0829 Supplier Cole-Parmer Supplier No. 0440665
Each compartment of this drawer rack holds two, 96-well boxes that easily can be retrieved from either side of rack. It is made of sturdy corrosion-resistant stainless steel for long-term use. Drawers slide out independently for quick access and storage of boxes.