Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Drawer Rack for Microtube Boxes with 1 1/2" Max Height, 5 x 5 Array
Catalog No. 501960825 Shop All Cole-Parmer Products
Encompass_Preferred
Each compartment of this drawer rack holds two microtube boxes that can be easily retrieved from either side of rack. It is made of sturdy corrosion-resistant stainless steel for long-term use
Catalog No. 50-196-0825 Supplier Cole-Parmer Supplier No. 0440597
Each compartment of this drawer rack holds two microtube boxes that can be easily retrieved from either side of rack. It is made of sturdy corrosion-resistant stainless steel for long-term use