Learn More
Medchemexpress LLC FzM1.8 50mg | 2204290-85-5 | 322.31 g/mol | C18H14N2O4 | 50 MG

Supplier: Medchemexpress LLC HY11716350MG
FzM1.8 is a small-molecule allosteric agonist of the Frizzled receptor FZD4 that activates the WNT/β-catenin signaling pathway by promoting TCF/LEF transcriptional activity in the absence of WNT ligand. The compound is provided as a research-grade, purified material with reported physicochemical and handling data suitable for in vitro and in vivo studies.
- Allosteric agonist of FZD4 that activates the WNT/β-catenin pathway.
- Suitable for in vitro and in vivo research applications.
- High reported purity (98.93%) for reproducible results.
- Molecular formula C18H14N2O4 and molecular weight 322.31 g/mol.
- Soluble in DMSO (≈83.33 mg/mL) and formulatable for in vivo dosing.
- Recommended storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.