Learn More
Research Products International Corp FlipTop FZR Box, for 81 Tubes, GRN, 5/cs
SDP
Catalog No. 502230164
Encompass
FlipTop Freezer Boxes with hinged lids are manufactured from recycled cardboard and are ideal for freezer and refrigerator use. Boxes contain cardboard cell partition to hold popular size micro-tubes (0.2 and 1.5 - 2.0 ml). Feature liquid resistant outer coating, alpha-numeric identification on top, side and bottom of boxes, including inside lid, grid markings and drain slots. Supplied 5 per case.
Hinged lid
Standard 5-1/4 x 5-1/2 x 2” size
Convenient
Catalog No. 50-223-0164 Supplier Research Products International Corp Supplier No. 249422G
FlipTop Freezer Boxes with hinged lids are manufactured from recycled cardboard and are ideal for freezer and refrigerator use. Boxes contain cardboard cell partition to hold popular size micro-tubes (0.2 and 1.5 - 2.0 ml). Feature liquid resistant outer coating, alpha-numeric identification on top, side and bottom of boxes, including inside lid, grid markings and drain slots. Supplied 5 per case.
Hinged lid
Standard 5-1/4 x 5-1/2 x 2” size
Convenient