Learn More
Research Products International Corp FlipTop FZR Boxes, for 100 Tubes, BL, 5/cs
SDP
Catalog No. 502230169
Encompass
FlipTop Freezer Boxes with hinged lids are manufactured from recycled cardboard and are ideal for freezer and refrigerator use. Boxes contain cardboard cell partition to hold popular size micro-tubes (0.2 and 1.5 - 2.0 ml). Feature liquid resistant outer coating, alpha-numeric identification on top, side and bottom of boxes, including inside lid, grid markings and drain slots. Supplied 5 per case.
Hinged lid
Standard 5-1/4 x 5-1/2 x 2” size
Convenient
Catalog No. 50-223-0169 Supplier Research Products International Corp Supplier No. 249423B
FlipTop Freezer Boxes with hinged lids are manufactured from recycled cardboard and are ideal for freezer and refrigerator use. Boxes contain cardboard cell partition to hold popular size micro-tubes (0.2 and 1.5 - 2.0 ml). Feature liquid resistant outer coating, alpha-numeric identification on top, side and bottom of boxes, including inside lid, grid markings and drain slots. Supplied 5 per case.
Hinged lid
Standard 5-1/4 x 5-1/2 x 2” size
Convenient