Learn More
Research Products International Corp FZR RK, Upright, for 15 & 50ml Tubes, 2x2 Array
SDP
Catalog No. 502229688
Encompass
Maximize your freezer storage with a freezer rack. The freezer rack can fit 5 3/4 inch square boxes with a 4 3/4 inch height. Box capacity is 4, in a 2 x 2 array. Boxes measuring 5 3/4 x 4 3/4 inch boxes are for storage of 15 and 50 ml tubes.
Catalog No. 50-222-9688 Supplier Research Products International Corp Supplier No. 247585
Maximize your freezer storage with a freezer rack. The freezer rack can fit 5 3/4 inch square boxes with a 4 3/4 inch height. Box capacity is 4, in a 2 x 2 array. Boxes measuring 5 3/4 x 4 3/4 inch boxes are for storage of 15 and 50 ml tubes.