Learn More
Aviva Systems Biology Corp FZR1 Antibody - N-terminal region (ARP51279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FZR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human FZR1. Alternate names: CDC20C, CDH1, FZR, FZR2, HCDH, HCDH1, KIAA1242ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-285-573