Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Sliding Tray Eco-Racks for Standard 3" Boxes, 4 x 3 Array
Catalog No. 501960805 Shop All Cole-Parmer Products
Encompass_Preferred
These top loading sliding tray eco-racks are a great economical alternative to the standing side access drawer racks. Trays slide out for easy retrieval of samples without having to remove the entire rack from the freezer.
Catalog No. 50-196-0805 Supplier Cole-Parmer Supplier No. 0440399
These top loading sliding tray eco-racks are a great economical alternative to the standing side access drawer racks. Trays slide out for easy retrieval of samples without having to remove the entire rack from the freezer.