Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 50-Cell Hinged Top Plastic Boxes, 6 x 4 Array
Catalog No. 501960552 Shop All Cole-Parmer Products
Encompass_Preferred
Conveniently store 50-cell hinged top plastic boxes that measure 2-1/4" x 3-3/8" x 5-9/16" with these racks. They come wiith locking rods to keep boxes securely in place. They racks are creafted of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0552 Supplier Cole-Parmer Supplier No. 0440508
Conveniently store 50-cell hinged top plastic boxes that measure 2-1/4" x 3-3/8" x 5-9/16" with these racks. They come wiith locking rods to keep boxes securely in place. They racks are creafted of sturdy corrosion-resistant stainless steel for long-term use.