Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Standard 2", 3" or 3.75" Boxes, 3 columns; 12 3/8"H
Catalog No. 501960821 Shop All Cole-Parmer Products
Encompass_Preferred
The open configuration of this rack lets you store a multiple combination of box sizes. Easily access boxes from either side of rack that is made of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0821 Supplier Cole-Parmer Supplier No. 0440488
The open configuration of this rack lets you store a multiple combination of box sizes. Easily access boxes from either side of rack that is made of sturdy corrosion-resistant stainless steel for long-term use.