Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for Standard 2", 3" or 3.75" Boxes, 5 columns; 8 1/4"H
Catalog No. 501960822 Shop All Cole-Parmer Products
Encompass_Preferred
The open configuration of this rack lets you store a multiple combination of box sizes. Easily access boxes from either side of rack that is made of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0822 Supplier Cole-Parmer Supplier No. 0440493
The open configuration of this rack lets you store a multiple combination of box sizes. Easily access boxes from either side of rack that is made of sturdy corrosion-resistant stainless steel for long-term use.