Learn More
Cole-Parmer Argos Technologies PolarSafe Upright Freezer Rack for 50-Cell 1.5 mL Boxes, 1 x 3 x 2 Array
Catalog No. 501960832 Shop All Cole-Parmer Products
Encompass_Preferred
Each compartment of this rack holds, two 50-cell 1.5 mL microtube storage boxes that can easily be retrieved from either side of the rack. It is made of sturdy corrosion-resistant stainless steel for long-term use.
Catalog No. 50-196-0832 Supplier Cole-Parmer Supplier No. 0440710
Each compartment of this rack holds, two 50-cell 1.5 mL microtube storage boxes that can easily be retrieved from either side of the rack. It is made of sturdy corrosion-resistant stainless steel for long-term use.