Learn More
Selleck Chemical LLC G-1 S0851-25MG
SDP
Catalog No. 5000042898
Encompass_Preferred
G-1 (LNS 8801) is a nonsteroidal and selective agonist of G protein-coupled receptor 30 (GPR30 G protein-coupled estrogen receptor 1 GPER) with Ki of 11 nM
Catalog No. 50-000-42898 Supplier Selleck Chemical LLC Supplier No. S085125MG
G-1 (LNS 8801) is a nonsteroidal and selective agonist of G protein-coupled receptor 30 (GPR30 G protein-coupled estrogen receptor 1 GPER) with Ki of 11 nM