Learn More
Medchemexpress LLC HY-107216 5mg Medchemexpress, G-1 CAS:881639-98-1 Purity:>98%
SDP
Catalog No. 501968372
Encompass_Preferred
Medchemexpress, HY-107216 5mg G-1 CAS:881639-98-1 G-1 is a nonsteroidal, high-affinity and selective agonist of GPR30 with a Ki of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8372 Supplier Medchemexpress LLC Supplier No. HY1072165MG
Medchemexpress, HY-107216 5mg G-1 CAS:881639-98-1 G-1 is a nonsteroidal, high-affinity and selective agonist of GPR30 with a Ki of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.