Learn More
Paul N Gardner Company G-200 CERTIFIED VISCOSITY STD
SDP
Catalog No. NC0155248
Encompass
G-200 Viscosity Standard packaged in 1 Pint, 0.47 Liter sealed, dark glass bottles. Label gives the Standard Name, Lot No., Termination Date, Viscosity in Centistokes at 25.00C (77.00F); traceable to NIST and comes with (M.S.D.S.)
Catalog No. NC0155248 Supplier Paul N Gardner Company Supplier No. VI3821
G-200 Viscosity Standard packaged in 1 Pint, 0.47 Liter sealed, dark glass bottles. Label gives the Standard Name, Lot No., Termination Date, Viscosity in Centistokes at 25.00C (77.00F); traceable to NIST and comes with (M.S.D.S.)