Learn More
Electron Microscopy Sciences Thompson Graticle G23, 21mm
SDP
Catalog No. 5029312
Encompass_Preferred
For counting particles in any of three areas of known size. The graticule is calibrated in the same manner as a normal eyepiece scale. The result is then used to calculate the area of any square.
Size: 21mm
Divisions: 10 mm, 7 mm and 4 mm squares
Catalog No. 50-293-12 Supplier Electron Microscopy Sciences Supplier No. 6809021
For counting particles in any of three areas of known size. The graticule is calibrated in the same manner as a normal eyepiece scale. The result is then used to calculate the area of any square.
Size: 21mm
Divisions: 10 mm, 7 mm and 4 mm squares