Learn More
Alkali Scientific G 418 Disulfate, 1 G
SDP
Catalog No. 5000659949
Encompass_Preferred
G418 Disulfate (1g) is an antibiotic commonly used in laboratory settings for cell culture selection. It is particularly effective in selecting for cells that have been transfected with a specific gene or plasmid that confers resistance to G418. G418 works by inhibiting protein synthesis in eukaryotic cells, leading to cell death unless the cells express the resistance gene. This makes G418 an essential tool for creating stable cell lines in molecular biology and biotechnology research. The 1g powder form is easy to use in laboratory protocols and can be dissolved in media for selection purposes, ensuring precise and controlled cell culture experiments.
Catalog No. 50-006-59949 Supplier Alkali Scientific Supplier No. GDI001CP
G418 Disulfate (1g) is an antibiotic commonly used in laboratory settings for cell culture selection. It is particularly effective in selecting for cells that have been transfected with a specific gene or plasmid that confers resistance to G418. G418 works by inhibiting protein synthesis in eukaryotic cells, leading to cell death unless the cells express the resistance gene. This makes G418 an essential tool for creating stable cell lines in molecular biology and biotechnology research. The 1g powder form is easy to use in laboratory protocols and can be dissolved in media for selection purposes, ensuring precise and controlled cell culture experiments.