Learn More
Alkali Scientific G 418 Disulfate, 10 G
SDP
Catalog No. 5000659726
Encompass_Preferred
G 418 Disulfate is a powerful antibiotic used in cell culture selection to maintain stable expression of transfected genes. In powder form, this 10g bottle is an essential tool for researchers working with mammalian and other eukaryotic cell lines. G418 is widely used for selecting and maintaining cells that have integrated the neomycin resistance gene, which is commonly used in gene expression and gene knockout studies. This reagent’s effectiveness in selecting resistant cells makes it crucial for a variety of genetic and pharmaceutical research applications, particularly in gene therapy, molecular biology, and drug discovery.
Catalog No. 50-006-59726 Supplier Alkali Scientific Supplier No. GDI002CP
G 418 Disulfate is a powerful antibiotic used in cell culture selection to maintain stable expression of transfected genes. In powder form, this 10g bottle is an essential tool for researchers working with mammalian and other eukaryotic cell lines. G418 is widely used for selecting and maintaining cells that have integrated the neomycin resistance gene, which is commonly used in gene expression and gene knockout studies. This reagent’s effectiveness in selecting resistant cells makes it crucial for a variety of genetic and pharmaceutical research applications, particularly in gene therapy, molecular biology, and drug discovery.