Learn More
Medchemexpress LLC HY-17561 1g , G-418 (disulfate) CAS:108321-42-2 Purity:>98%
SDP
Catalog No. 502259204
Encompass_Preferred
HY-17561 1g G-418 (disulfate) CAS: 108321-42-2 G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9204 Supplier Medchemexpress LLC Supplier No. HY175611G
HY-17561 1g G-418 (disulfate) CAS: 108321-42-2 G-418 disulfate (Geneticin sulfate), is an aminoglycoside antibiotic, inhibits protein synthesis in eukaryotes and prokaryotes. G-418 disulfate is commonly used as a selective agent for eukaryotic cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.