Learn More
Alkali Scientific G 418 Disulfate, 25 G
SDP
Catalog No. 5000660533
Encompass_Preferred
G 418 Disulfate in a 25g powder form is a highly effective antibiotic used for cell culture selection, particularly in mammalian cells. This reagent helps in the selection of cells that have successfully integrated the neomycin resistance gene, commonly used in the context of transfection experiments. The high concentration of G418 in this 25g format makes it a cost-effective solution for large-scale cell culture applications. Researchers use this antibiotic to ensure that only cells that have incorporated the transfected gene will survive and proliferate, enabling more efficient genetic manipulation and screening processes.
Catalog No. 50-006-60533 Supplier Alkali Scientific Supplier No. GDI003CP
G 418 Disulfate in a 25g powder form is a highly effective antibiotic used for cell culture selection, particularly in mammalian cells. This reagent helps in the selection of cells that have successfully integrated the neomycin resistance gene, commonly used in the context of transfection experiments. The high concentration of G418 in this 25g format makes it a cost-effective solution for large-scale cell culture applications. Researchers use this antibiotic to ensure that only cells that have incorporated the transfected gene will survive and proliferate, enabling more efficient genetic manipulation and screening processes.