Learn More
Alkali Scientific G 418 Disulfate, 5 G
SDP
Catalog No. 5000661173
Encompass_Preferred
G 418 Disulfate is available in a 5g powder form for cell culture selection, ensuring the survival of only those cells that have successfully incorporated the neomycin resistance gene. G418 is commonly used in mammalian and other eukaryotic cell cultures, particularly in genetic manipulation studies such as gene expression and gene knockout research. This high-efficiency antibiotic selects for transformed cells, making it a crucial reagent for genetic studies and pharmaceutical research. This 5g format is perfect for smaller-scale experiments or as a supplement to larger quantities for long-term use in research.
Catalog No. 50-006-61173 Supplier Alkali Scientific Supplier No. GDI004CP
G 418 Disulfate is available in a 5g powder form for cell culture selection, ensuring the survival of only those cells that have successfully incorporated the neomycin resistance gene. G418 is commonly used in mammalian and other eukaryotic cell cultures, particularly in genetic manipulation studies such as gene expression and gene knockout research. This high-efficiency antibiotic selects for transformed cells, making it a crucial reagent for genetic studies and pharmaceutical research. This 5g format is perfect for smaller-scale experiments or as a supplement to larger quantities for long-term use in research.