Learn More
Alkali Scientific G 418 Disulfate 50 Mg/mL Solution, 20 Milliliters
SDP
Catalog No. 5000660228
Encompass_Preferred
G 418 Disulfate 50 mg/mL solution in a 20mL vial is used for cell culture selection, ensuring that only transfected cells that have integrated the neomycin resistance gene can survive. This highly concentrated antibiotic solution is widely used in research involving mammalian cells, especially in gene expression and gene knockout experiments. The 50 mg/mL concentration provides an effective selection tool for large-scale experiments, and the liquid form allows for easy, accurate dosing. G418 is essential for maintaining stable gene expression and for studying the effects of specific genes in various cell lines.
Catalog No. 50-006-60228 Supplier Alkali Scientific Supplier No. GDI006CP
G 418 Disulfate 50 mg/mL solution in a 20mL vial is used for cell culture selection, ensuring that only transfected cells that have integrated the neomycin resistance gene can survive. This highly concentrated antibiotic solution is widely used in research involving mammalian cells, especially in gene expression and gene knockout experiments. The 50 mg/mL concentration provides an effective selection tool for large-scale experiments, and the liquid form allows for easy, accurate dosing. G418 is essential for maintaining stable gene expression and for studying the effects of specific genes in various cell lines.