Learn More
Medchemexpress LLC HY-17561 5g , G-418 (disulfate) Geneticin sulfate;Antibiotic G-418 sulfate CAS:108321-42-2 Purity:98%
SDP
Catalog No. 501492177
Encompass_Preferred
Medchemexpress, HY-17561 5g G-418 (disulfate) Geneticin sulfate;Antibiotic G-418 sulfate CAS:108321-42-2 Formula:C20H44N4O18S2 Purity:98% G-418 (disulfate) is an aminoglycoside antibiotic similar in structure to gentamicin B1, which blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2177 Supplier Medchemexpress LLC Supplier No. HY175615G
Medchemexpress, HY-17561 5g G-418 (disulfate) Geneticin sulfate;Antibiotic G-418 sulfate CAS:108321-42-2 Formula:C20H44N4O18S2 Purity:98% G-418 (disulfate) is an aminoglycoside antibiotic similar in structure to gentamicin B1, which blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.