Learn More
Crescent Chemical Co Inc G-418 SOLUTION
SDP
Catalog No. 50249147
Encompass_Preferred
G-418 solution solution Formulated to contain 50 mg/ml G 418 base in deionized water. 50 mg G 418 base are approx. 70 mg G 418 sulfate (based on dry weight). Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog Number - 47995.01
Catalog No. 50-249-147 Supplier Crescent Chemical Co Inc Supplier No. SE47995.01
G-418 solution solution Formulated to contain 50 mg/ml G 418 base in deionized water. 50 mg G 418 base are approx. 70 mg G 418 sulfate (based on dry weight). Size - 20ML Storage Conditions - -15 °C TO -25 °C Catalog Number - 47995.01