Learn More
Gold Biotechnology Inc G-418 Sulfate 5 g
SDP
Catalog No. 501532787
Encompass_Preferred
G-418 sulfate, also known as geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1. It is commonly used in plant and mammalian cell culture as a selectable marker. It can be used to select for cells transformed with an iNOS promoter construct or neomycin resistance gene.
Catalog No. 50-153-2787 Supplier Gold Biotechnology Inc Supplier No. G4185
G-418 sulfate, also known as geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1. It is commonly used in plant and mammalian cell culture as a selectable marker. It can be used to select for cells transformed with an iNOS promoter construct or neomycin resistance gene.