Learn More
Crescent Chemical Co Inc G 418 SULFATE
SDP
Supplier:  Crescent Chemical Co Inc SE49418.03
Encompass_Preferred
G 418 sulfate Aminoglycoside antibiotic from Micromonospora rhodorangea. Used for the selection and maintenance of eukaryotic cells expressing the neomycin resistance gene (neo). G418 blocks polypeptide synthesis by inhibiting the elongation step. It i
Catalog No. 50-249-194