Learn More
Gold Biotechnology Inc G-418 Sulfate 1 g
SDP
Catalog No. 501532784
Encompass_Preferred
G-418 sulfate, also known as geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1. It is commonly used in plant and mammalian cell culture as a selectable marker. It can be used to select for cells transformed with an iNOS promoter construct or neomycin resistance gene.
Catalog No. 50-153-2784 Supplier Gold Biotechnology Inc Supplier No. G4181
G-418 sulfate, also known as geneticin, is an aminoglycoside antibiotic similar in structure to gentamicin B1. It is commonly used in plant and mammalian cell culture as a selectable marker. It can be used to select for cells transformed with an iNOS promoter construct or neomycin resistance gene.