Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ G-5 Supplement (100X)

Catalog No. 17503012
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-503-012 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-503-012 Supplier Gibco™ Supplier No. 17503012
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

G-5 Supplement (100X)

G-5 Supplement is a chemically defined, serum-free supplement based on Bottenstein’s G-5 formulation. G-5 Supplement is intended for use with Neurobasal™ media.

G-5 Supplement is recommended for growth and expression of glial cells (normal and tumor) of astrocytic phenotype (astrocytes). G-5 Supplement can be used as a substitute for Bottenstein’s G-2 formulation. G-5 Supplement is provided as a 100X concentrate.

G-5 Supplement may be used with Neurobasal™ media, DMEM, DMEM⁄F-12, or other basal media..

Learn more about G-5 Supplement formulation.

Learn more about neural cell culture products from Invitrogen at www.invitrogen.com/neuralculture.

For Research Use Only. Not intended for any animal or human therapeutic of diagnostic use.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.

Cell Type Glial Cells
Classification Serum-free
Concentration 100 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
Quantity 1 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.